ENA413166196 Enamine

ethyl 2-bromo-5-chloropyridine-4-carboxylate

  • MDL number MFCD14698305
  • Cas Number : 1214357-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140864
Availability details for
Quantity
  • Availability
  • Add to Cart