ENA413166198 Enamine

tert-butyl N-[1-cyano-3-(propan-2-yloxy)propyl]carbamate

  • MDL number MFCD27500704
  • Cas Number : 1797386-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140872
Availability details for
Quantity
  • Availability
  • Add to Cart