ENA413166200 Enamine

2-(2-iodoethoxy)propane

  • MDL number MFCD20414987
  • Cas Number : 318958-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140875

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140875
Availability details for
Quantity
  • Availability
  • Add to Cart