ENA413166204 Enamine

2-amino-3-(2-chloro-6-fluorophenyl)propanoic acid

  • MDL number MFCD04116005
  • Cas Number : 603940-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140886
Availability details for
Quantity
  • Availability
  • Add to Cart