ENA413166214 Enamine

2-fluoro-5-methyl-4-nitrophenol

  • MDL number MFCD22042960
  • Cas Number : 63762-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140907
Availability details for
Quantity
  • Availability
  • Add to Cart