ENA413166219 Enamine

1-cyclobutylprop-2-yn-1-one

  • MDL number MFCD21263108
  • Cas Number : 1466042-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140937
Availability details for
Quantity
  • Availability
  • Add to Cart