ENA413166220 Enamine

1-iodo-4-methoxy-2-methylbenzene

  • MDL number MFCD11101045
  • Cas Number : 63452-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140939
Availability details for
Quantity
  • Availability
  • Add to Cart