ENA413166227 Enamine

2-[(2-ethylcyclohexyl)oxy]ethan-1-ol

  • MDL number MFCD16102292
  • Cas Number : 1250390-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140972

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140972
Availability details for
Quantity
  • Availability
  • Add to Cart