ENA413166232 Enamine

(2-amino-1-phenylpropyl)dimethylamine

  • MDL number MFCD16061675
  • Cas Number : 104177-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140979

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140979
Availability details for
Quantity
  • Availability
  • Add to Cart