ENA413166234 Enamine

4-(3-aminopropoxy)pyridine

  • MDL number MFCD16714231
  • Cas Number : 155967-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140982

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140982
Availability details for
Quantity
  • Availability
  • Add to Cart