ENA413166239 Enamine

2-methylpyrrolidine-1-sulfonyl chloride

  • MDL number MFCD12547693
  • Cas Number : 1184477-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140997
Availability details for
Quantity
  • Availability
  • Add to Cart