ENA413166245 Enamine

{1-[2-(benzyloxy)ethyl]cyclopropyl}methanamine

  • MDL number MFCD26936154
  • Cas Number : 1797324-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141008

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141008
Availability details for
Quantity
  • Availability
  • Add to Cart