ENA413166246 Enamine

2-chloro-4-(5-phenyl-1H-1,2,4-triazol-3-yl)aniline

  • MDL number MFCD21384960
  • Cas Number : 1485415-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141010
Availability details for
Quantity
  • Availability
  • Add to Cart