ENA413166247 Enamine

4-methyl-2-(1-methyl-1H-pyrazol-4-yl)-1,3-oxazole-5-carboxylic acid

  • MDL number MFCD21291568
  • Cas Number : 1498817-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233...235
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141013
Availability details for
Quantity
  • Availability
  • Add to Cart