ENA413166255 Enamine

1-cyclopropyl-3-(prop-2-yn-1-yl)urea

  • MDL number MFCD16664559
  • Cas Number : 1249183-21-8
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141034
Availability details for
Quantity
  • Availability
  • Add to Cart