ENA413166261 Enamine

(2-amino-2-phenylethyl)dimethylamine dihydrochloride

  • MDL number MFCD11974392
  • Cas Number : 31788-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141053

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141053
Availability details for
Quantity
  • Availability
  • Add to Cart