ENA413166269 Enamine

3-(ethylsulfanyl)benzaldehyde

  • MDL number MFCD21760494
  • Cas Number : 87425-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141080

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141080
Availability details for
Quantity
  • Availability
  • Add to Cart