ENA413166270 Enamine

6-(difluoromethoxy)-1-benzofuran-2-carboxylic acid

  • MDL number MFCD26936161
  • Cas Number : 1803591-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141081
Availability details for
Quantity
  • Availability
  • Add to Cart