ENA413166271 Enamine

1-(3-hydroxythiophen-2-yl)ethan-1-one

  • MDL number MFCD07787415
  • Cas Number : 5556-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141082

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141082
Availability details for
Quantity
  • Availability
  • Add to Cart