ENA413166275 Enamine

N1-(1-methanesulfonylpiperidin-4-yl)-4-methylbenzene-1,3-diamine

  • MDL number MFCD17142511
  • Cas Number : 1409216-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141088
Availability details for
Quantity
  • Availability
  • Add to Cart