ENA413166276 Enamine

2-(3-chloro-2-fluorophenyl)-2-methylpropanenitrile

  • MDL number MFCD11036558
  • Cas Number : 1260853-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141089
Availability details for
Quantity
  • Availability
  • Add to Cart