ENA413166279 Enamine

6-cyclopropyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid

  • MDL number MFCD22065611
  • Cas Number : 150190-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141093

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141093
Availability details for
Quantity
  • Availability
  • Add to Cart