ENA413166283 Enamine

3-chloro-N-[(4-nitrophenyl)methyl]propane-1-sulfonamide

  • MDL number MFCD13158046
  • Cas Number : 258856-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141119

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141119
Availability details for
Quantity
  • Availability
  • Add to Cart