ENA413166287 Enamine

tert-butyl N-[2-(prop-2-enamido)ethyl]carbamate

  • MDL number MFCD24468676
  • Cas Number : 165196-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141144

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141144
Availability details for
Quantity
  • Availability
  • Add to Cart