ENA413166288 Enamine

methyl 4-amino-2-(dimethylamino)pyrimidine-5-carboxylate

  • MDL number MFCD26936168
  • Cas Number : 1600893-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141145

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141145
Availability details for
Quantity
  • Availability
  • Add to Cart