ENA413166291 Enamine

bis(1-(2,2,2-trifluoroethyl)guanidine); sulfuric acid

  • MDL number MFCD26936069
  • Cas Number : 551919-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140235

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-140235
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart