ENA413166296 Enamine

4-(furan-2-yl)pyridine

  • MDL number MFCD27500707
  • Cas Number : 55484-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141191

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141191
Availability details for
Quantity
  • Availability
  • Add to Cart