ENA413166302 Enamine

5-cyclopropyl-1-(propan-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD06805483
  • Cas Number : 1006496-39-4
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-cyclopropyl-1-isopropyl-1H-pyrazol-4-amine
Inchi Code 1S/C9H15N3/c1-6(2)12-9(7-3-4-7)8(10)5-11-12/h5-7H,3-4,10H2,1-2H3
Inchi Key LVESYDJCYODKFP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-141222

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-141222
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart