ENA413166306 Enamine

(1R)-1-(oxan-4-yl)ethan-1-ol

  • MDL number MFCD21188408
  • Cas Number : 1568198-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141228
Availability details for
Quantity
  • Availability
  • Add to Cart