ENA413166322 Enamine

3-(4-ethylpiperazin-1-yl)aniline

  • MDL number MFCD10032982
  • Cas Number : 314061-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141266
Availability details for
Quantity
  • Availability
  • Add to Cart