ENA413166325 Enamine

2,4,6-trimethoxybenzene-1-sulfonyl chloride

  • MDL number MFCD08706423
  • Cas Number : 52499-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-136
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141273
Availability details for
Quantity
  • Availability
  • Add to Cart