ENA413166328 Enamine

3-bromo-8-methoxyquinoline

  • MDL number MFCD22690585
  • Cas Number : 103030-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141288

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141288
Availability details for
Quantity
  • Availability
  • Add to Cart