ENA413166329 Enamine

ethyl 2-ethyl-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD08669883
  • Cas Number : 354587-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141289
Availability details for
Quantity
  • Availability
  • Add to Cart