ENA413166332 Enamine

ethyl N-[2-(trifluoromethyl)phenoxy]ethanecarboximidate

  • MDL number MFCD26936176
  • Cas Number : 849048-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141300

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141300
Availability details for
Quantity
  • Availability
  • Add to Cart