ENA413166335 Enamine

5-chloro-2-{[(6-methylpyridin-2-yl)methyl]amino}benzoic acid

  • MDL number MFCD17957912
  • Cas Number : 1283531-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140295
Availability details for
Quantity
  • Availability
  • Add to Cart