ENA413166338 Enamine

tert-butyl N-(4-methyl-2-propyl-1,3-thiazol-5-yl)carbamate

  • MDL number MFCD26936179
  • Cas Number : 1797012-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141329
Availability details for
Quantity
  • Availability
  • Add to Cart