ENA413166340 Enamine

6-(tert-butylamino)hexanoic acid

  • MDL number MFCD20293006
  • Cas Number : 1339161-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141334

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141334
Availability details for
Quantity
  • Availability
  • Add to Cart