ENA413166342 Enamine

5-[(4-bromo-1H-pyrazol-1-yl)methyl]-2-chloro-1,3-thiazole

  • MDL number MFCD23765893
  • Cas Number : 1525598-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141348
Availability details for
Quantity
  • Availability
  • Add to Cart