ENA413166345 Enamine

2-methylbutanethioamide

  • MDL number MFCD11168463
  • Cas Number : 88512-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140307

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140307
Availability details for
Quantity
  • Availability
  • Add to Cart