ENA413166351 Enamine

3-amino-4-(trifluoromethoxy)benzoic acid

  • MDL number MFCD18390052
  • Cas Number : 656-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139652

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139652
Availability details for
Quantity
  • Availability
  • Add to Cart