ENA413166354 Enamine

2-methyl-6-(methylsulfanyl)aniline

  • MDL number MFCD13677164
  • Cas Number : 100305-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-141366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141366
Availability details for
Quantity
  • Availability
  • Add to Cart