ENA413166356 Enamine

4-(5-amino-4H-1,2,4-triazol-3-yl)-2-chlorophenol

  • MDL number MFCD09806275
  • Cas Number : 1016534-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 273...275
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141371

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141371
Availability details for
Quantity
  • Availability
  • Add to Cart