ENA413166361 Enamine

[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]methanol

  • MDL number MFCD12137986
  • Cas Number : 1155531-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141377

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141377
Availability details for
Quantity
  • Availability
  • Add to Cart