ENA413166365 Enamine

[(3-chloro-4-methylphenyl)carbamoyl](phenyl)methyl 4-(carbamoylamino)benzoate

  • MDL number MFCD04626146
  • Cas Number : 562860-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-141398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141398
Availability details for
Quantity
  • Availability
  • Add to Cart