ENA413166369 Enamine

{[(2,2-dimethoxyethyl)sulfanyl]methyl}benzene

  • MDL number MFCD21728552
  • Cas Number : 6304-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141416
Availability details for
Quantity
  • Availability
  • Add to Cart