ENA413166371 Enamine

N-(6-amino-1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)formamide

  • MDL number MFCD00068888
  • Cas Number : 7597-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-141445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141445
Availability details for
Quantity
  • Availability
  • Add to Cart