ENA413166381 Enamine

3-(4-fluorophenyl)-2-methylpropan-1-ol

  • MDL number MFCD20386076
  • Cas Number : 1186584-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141456

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141456
Availability details for
Quantity
  • Availability
  • Add to Cart