ENA413166385 Enamine

3-bromo-1-[2-(trifluoromethyl)phenyl]pyrrolidin-2-one

  • MDL number MFCD19527249
  • Cas Number : 1412417-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141463
Availability details for
Quantity
  • Availability
  • Add to Cart