ENA413166393 Enamine

2-(3-chloro-2-fluorophenyl)propan-2-amine hydrochloride

  • MDL number MFCD26936191
  • Cas Number : 1803591-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141478
Availability details for
Quantity
  • Availability
  • Add to Cart