ENA413166396 Enamine

methyl 2-amino-3-(benzyloxy)benzoate

  • MDL number MFCD23675453
  • Cas Number : 533897-94-8
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-141481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141481
Availability details for
Quantity
  • Availability
  • Add to Cart